Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022603.1 | NP_072125.1 | 1 | 1031 | 116 | |
XM_006251078.2 | XP_006251140.1 | 2 | 1163 | 116 | ||
XM_006251079.2 | XP_006251141.1 | 2 | 1085 | 116 | ||
GenBank mRNA | AF142758.1 | - | 1 | 1031 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |