Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022605.1 | NP_072127.1 | 3 - 4 | 544 | 85 | |
GenBank mRNA | AF184967.1 | - | 3 - 4 | 544 | 85 | |
AF359508.1 | - | 3 - 4 | 571 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |