Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pth2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031089.2 | NP_112351.1 | 10 - 11 | 1281 | 164 | |
GenBank mRNA | U55836.1 | - | 10 - 11 | 1281 | 164 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |