Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053358.1 | NP_445810.1 | 8 - 9 | 911 | 142 | |
XM_006238533.2 | XP_006238595.1 | 9 - 10 | 1026 | 142 | ||
XM_006238535.2 | XP_006238597.1 | 9 - 10 | 1024 | 142 | ||
XM_006238537.2 | XP_006238599.1 | 8 - 9 | 942 | 142 | ||
XM_008763904.1 | XP_008762126.1 | 9 - 10 | 2677 | 142 | ||
XM_008763906.1 | XP_008762128.1 | 9 - 10 | 2674 | 142 | ||
XM_008763908.1 | XP_008762130.1 | 8 - 9 | 2593 | 142 | ||
XM_008763909.1 | XP_008762131.1 | 8 - 9 | 2590 | 142 | ||
GenBank mRNA | AF121893.1 | - | 8 - 9 | 911 | 142 | |
AY724490.1 | - | 9 - 10 | 692 | 142 | ||
BC107666.1 | - | 8 - 9 | 895 | 7936459 | 142 | |
FQ230168.1 | - | 3 - 4 | 189 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |