Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057188.1 | NP_476536.1 | 1 - 2 | 168 | 73 | |
GenBank mRNA | AF090867.1 | - | 1 - 2 | 168 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |