Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134468.1 | NP_604463.1 | 3 - 4 | 334 | 83 | |
XM_006236999.2 | XP_006237061.1 | 4 - 5 | 1003 | 83 | ||
XM_006237000.2 | XP_006237062.1 | 4 - 5 | 999 | 83 | ||
XM_006237001.2 | XP_006237063.1 | 3 - 4 | 866 | 83 | ||
XM_006237002.2 | XP_006237064.1 | 3 - 4 | 793 | 83 | ||
XM_006237003.2 | XP_006237065.1 | 3 - 4 | 792 | 83 | ||
GenBank mRNA | BC071177.1 | - | 3 - 4 | 269 | 7098592 | 83 |
FQ213485.1 | - | 297 | 83 | |||
L24907.1 | - | 3 - 4 | 308 | 83 | ||
L26288.1 | - | 3 - 4 | 334 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |