Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153298.1 | NP_695210.1 | 11 - 12 | 786 | 63 | |
XM_008763051.1 | XP_008761273.1 | 10 - 11 | 1353 | 63 | ||
GenBank mRNA | AB056724.1 | - | 11 - 12 | 803 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |