Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Has3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172319.1 | NP_758822.1 | 3 - 4 | 910 | 73 | |
XM_006255561.2 | XP_006255623.1 | 3 - 4 | 2130 | 73 | ||
GenBank mRNA | AB097569.1 | - | 3 - 4 | 910 | 73 | |
AF543196.1 | - | 2 - 3 | 368 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |