Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178333.1 | NP_848016.1 | 11 - 12 | 1399 | 88 | |
XM_008773069.1 | XP_008771291.1 | 13 - 14 | 2729 | 88 | ||
XM_008773070.1 | XP_008771292.1 | 13 - 14 | 2720 | 88 | ||
GenBank mRNA | AB098162.1 | - | 11 - 12 | 1399 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |