Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198789.2 | NP_942084.1 | 8 - 9 | 1063 | 99 | |
XM_006245000.2 | XP_006245062.2 | 8 - 9 | 1062 | 99 | ||
GenBank mRNA | BC061580.1 | - | 8 - 9 | 1067 | 6890414 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |