Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012776.1 | NP_036908.1 | 7 - 8 | 789 | 80 | |
GenBank mRNA | M87854.1 | - | 7 - 8 | 789 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |