Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017325.1 | NP_059021.1 | 2 - 3 | 829 | 74 | |
XM_008768598.1 | XP_008766820.1 | 2 - 3 | 2631 | 74 | ||
XM_008768599.1 | XP_008766821.1 | 2 - 3 | 2631 | 74 | ||
GenBank mRNA | L35271.1 | - | 2 - 3 | 829 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |