Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019175.1 | NP_062048.1 | 4 - 5 | 409 | 67 | |
XM_006228982.2 | XP_006229044.1 | 4 - 5 | 516 | 67 | ||
GenBank mRNA | AF016269.1 | - | 4 - 5 | 409 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Rattus norvegicus Probe spans exons |