Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019259.2 | NP_062132.2 | 2 - 3 | 384 | 82 | |
GenBank mRNA | AJ001102.1 | - | 2 - 3 | 425 | 82 | |
BC070510.1 | - | 2 - 3 | 384 | 7100505 | 82 | |
FQ212622.1 | - | 2 - 3 | 409 | 82 | ||
FQ214537.1 | - | 2 - 3 | 434 | 82 | ||
FQ219796.1 | - | 2 - 3 | 407 | 82 | ||
FQ220621.1 | - | 2 - 3 | 427 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |