Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021742.1 | NP_068510.1 | 4 - 5 | 472 | 60 | |
XM_006249928.2 | XP_006249990.1 | 3 - 4 | 580 | 60 | ||
XM_006249929.2 | XP_006249991.1 | 2 - 3 | 320 | 60 | ||
GenBank mRNA | AB012960.1 | - | 4 - 5 | 472 | 60 | |
AB012961.1 | - | 3 - 4 | 409 | 60 | ||
U47280.1 | - | 1 - 2 | 126 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor Rattus norvegicus Probe spans exons |