Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031010.2 | NP_112272.2 | 11 - 12 | 1550 | 72 | |
XM_006246823.2 | XP_006246885.1 | 13 - 14 | 1831 | 72 | ||
XM_006246824.2 | XP_006246886.1 | 12 - 13 | 1726 | 72 | ||
GenBank mRNA | L06040.1 | - | 11 - 12 | 1563 | 72 | |
S69383.1 | - | 11 - 12 | 1550 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |