Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053568.1 | NP_446020.1 | 5 - 6 | 537 | 59 | |
XM_006247935.2 | XP_006247997.1 | 5 - 6 | 563 | 59 | ||
XM_006247936.2 | XP_006247998.1 | 5 - 6 | 860 | 59 | ||
GenBank mRNA | AF080568.1 | - | 5 - 6 | 537 | 59 | |
BC078772.1 | - | 5 - 6 | 548 | 7111560 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |