Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133424.2 | NP_596915.1 | 9 - 10 | 933 | 89 | |
GenBank mRNA | AF450248.1 | - | 9 - 10 | 934 | 89 | |
AY096238.1 | - | 9 - 10 | 858 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |