Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133580.1 | NP_598264.1 | 7 - 8 | 501 | 152 | |
GenBank mRNA | BC061984.1 | - | 8 - 9 | 809 | 5599180 | 152 |
U18771.1 | - | 7 - 8 | 501 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |