Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153629.1 | NP_705893.1 | 9 - 10 | 1359 | 73 | |
XM_006246232.1 | XP_006246294.1 | 9 - 10 | 1403 | 73 | ||
GenBank mRNA | AF077354.1 | - | 9 - 10 | 1359 | 73 | |
FQ222433.1 | - | 10 - 11 | 1567 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |