Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153730.2 | NP_714952.1 | 6 - 7 | 952 | 67 | |
XM_006249843.2 | XP_006249905.1 | 7 - 8 | 1101 | 67 | ||
XM_006249844.2 | XP_006249906.1 | 7 - 8 | 1375 | 67 | ||
XM_006249845.1 | XP_006249907.1 | 7 - 8 | 1081 | 67 | ||
XM_006249846.2 | XP_006249908.1 | 8 - 9 | 1167 | 67 | ||
XM_008769539.1 | XP_008767761.1 | 7 - 8 | 1140 | 67 | ||
XM_008769540.1 | XP_008767762.1 | 8 - 9 | 995 | 67 | ||
XM_008769541.1 | XP_008767763.1 | 7 - 8 | 885 | 67 | ||
GenBank mRNA | AJ133126.1 | - | 6 - 7 | 994 | 67 | |
BC086983.1 | - | 7 - 8 | 1082 | 7133597 | 67 | |
FQ210179.1 | - | 8 - 9 | 1151 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |