Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198748.1 | NP_942043.1 | 10 - 11 | 1409 | 76 | |
XM_008764644.1 | XP_008762866.1 | 10 - 11 | 1747 | 76 | ||
XM_008764645.1 | XP_008762867.1 | 8 - 9 | 1222 | 76 | ||
GenBank mRNA | AY324394.1 | - | 10 - 11 | 1409 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |