Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198749.2 | NP_942044.1 | 3 - 4 | 622 | 80 | |
GenBank mRNA | BC094954.1 | - | 3 - 4 | 622 | 7316003 | 80 |
M83679.1 | - | 3 - 4 | 466 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |