Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198757.2 | NP_942052.1 | 6 - 7 | 847 | 114 | |
XM_008768010.1 | XP_008766232.1 | 5 - 6 | 762 | 114 | ||
XM_008768011.1 | XP_008766233.1 | 5 - 6 | 760 | 114 | ||
XM_008768012.1 | XP_008766234.1 | 5 - 6 | 643 | 114 | ||
XM_008768013.1 | XP_008766235.1 | 5 - 6 | 643 | 114 | ||
XM_008768014.1 | XP_008766236.1 | 5 - 6 | 597 | 114 | ||
XM_008768015.1 | XP_008766237.1 | 5 - 6 | 665 | 114 | ||
GenBank mRNA | AB106282.1 | - | 5 - 6 | 617 | 114 | |
BC082014.1 | - | 6 - 7 | 847 | 7132082 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |