Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198757.2 | NP_942052.1 | 7 - 8 | 914 | 129 | |
XM_008768010.1 | XP_008766232.1 | 6 - 7 | 829 | 129 | ||
XM_008768011.1 | XP_008766233.1 | 6 - 7 | 827 | 129 | ||
XM_008768012.1 | XP_008766234.1 | 6 - 7 | 710 | 129 | ||
XM_008768013.1 | XP_008766235.1 | 6 - 7 | 710 | 129 | ||
XM_008768014.1 | XP_008766236.1 | 6 - 7 | 664 | 129 | ||
XM_008768015.1 | XP_008766237.1 | 6 - 7 | 732 | 129 | ||
GenBank mRNA | AB106282.1 | - | 6 - 7 | 684 | 129 | |
BC082014.1 | - | 7 - 8 | 914 | 7132082 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |