Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201562.3 | NP_963856.1 | 8 - 9 | 1277 | 133 | |
GenBank mRNA | AY279211.1 | - | 8 - 9 | 1277 | 133 | |
BC129079.1 | - | 8 - 9 | 1392 | 8370402 | 133 | |
FQ215784.1 | - | 5 - 6 | 529 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |