Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scnn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012648.1 | NP_036780.1 | 8 - 9 | 1318 | 112 | |
XM_008759682.1 | XP_008757904.1 | 8 - 9 | 1353 | 112 | ||
XM_008759683.1 | XP_008757905.1 | 9 - 10 | 1563 | 112 | ||
XM_008759684.1 | XP_008757906.1 | 8 - 9 | 1395 | 112 | ||
XM_008759685.1 | XP_008757907.1 | 8 - 9 | 1353 | 112 | ||
XM_008759686.1 | XP_008757908.1 | 8 - 9 | 1353 | 112 | ||
GenBank mRNA | EU627516.1 | - | 7 - 8 | 1263 | 112 | |
U35174.1 | - | 7 - 8 | 1273 | 112 | ||
U35175.1 | - | 7 - 8 | 1273 | 112 | ||
X77932.1 | - | 8 - 9 | 1335 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons |