Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019244.2 | NP_062117.2 | 7 - 8 | 897 | 98 | |
GenBank mRNA | M93271.1 | - | 762 | 98 | ||
U27456.1 | - | 6 - 7 | 551 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |