Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF136401.1 | - | 67 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus |