Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eomes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766711.1 | XP_008764933.1 | 5 - 6 | 2003 | 73 | |
GenBank mRNA | AY457971.1 | - | 2 - 3 | 277 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |