Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762110.1 | XP_008760332.1 | 11 - 12 | 812 | 119 | |
XM_575205.5 | XP_575205.2 | 11 - 12 | 756 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |