Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762110.1 | XP_008760332.1 | 2 - 3 | 124 | 102 | |
XM_575205.5 | XP_575205.2 | 2 - 3 | 68 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Probe spans exons |