Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762236.1 | XP_008760458.1 | 8 - 9 | 2667 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |