Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236119.2 | XP_006236181.1 | 2 - 3 | 1591 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |