Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024322.1 | NP_001019493.1 | 11 - 12 | 1488 | 73 | |
XM_006236160.2 | XP_006236222.1 | 12 - 13 | 1580 | 73 | ||
XM_006236161.2 | XP_006236223.1 | 13 - 14 | 1802 | 73 | ||
XM_008762809.1 | XP_008761031.1 | 13 - 14 | 1984 | 73 | ||
XM_008762810.1 | XP_008761032.1 | 13 - 14 | 1929 | 73 | ||
XM_008762811.1 | XP_008761033.1 | 11 - 12 | 1533 | 73 | ||
GenBank mRNA | BC085834.1 | - | 11 - 12 | 1488 | 7134840 | 73 |
BC090024.1 | - | 6 - 7 | 904 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |