Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236158.2 | XP_006236220.1 | 2 - 3 | 464 | 64 | |
XM_008762831.1 | XP_008761053.1 | 2 - 3 | 620 | 64 | ||
XM_008762832.1 | XP_008761054.1 | 2 - 3 | 421 | 64 | ||
XM_008762833.1 | XP_008761055.1 | 2 - 3 | 642 | 64 | ||
GenBank mRNA | BC097405.1 | - | 2 - 3 | 176 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |