Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236158.2 | XP_006236220.1 | 7 - 8 | 1223 | 73 | |
XM_008762831.1 | XP_008761053.1 | 7 - 8 | 1379 | 73 | ||
XM_008762832.1 | XP_008761054.1 | 7 - 8 | 1180 | 73 | ||
XM_008762833.1 | XP_008761055.1 | 7 - 8 | 1401 | 73 | ||
GenBank mRNA | BC097405.1 | - | 7 - 8 | 935 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |