Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107853.1 | NP_001101323.1 | 2 - 3 | 489 | 136 | |
XM_006236319.2 | XP_006236381.1 | 2 - 3 | 921 | 136 | ||
XM_006236321.2 | XP_006236383.1 | 2 - 3 | 559 | 136 | ||
XM_008762780.1 | XP_008761002.1 | 3 - 4 | 428 | 136 | ||
GenBank mRNA | BC166855.1 | - | 2 - 3 | 510 | 9099059 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |