Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011952.1 | NP_001011952.1 | 3 - 4 | 803 | 95 | |
XM_006233356.2 | XP_006233418.1 | 4 - 5 | 842 | 95 | ||
XM_006233357.2 | XP_006233419.1 | 4 - 5 | 1062 | 95 | ||
GenBank mRNA | BC089844.1 | - | 3 - 4 | 803 | 7368889 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |