Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233199.2 | XP_006233261.1 | 4 - 5 | 523 | 96 | |
XM_006233200.2 | XP_006233262.1 | 4 - 5 | 523 | 96 | ||
XM_006233204.2 | XP_006233266.1 | 4 - 5 | 1886 | 96 | ||
XM_008761446.1 | XP_008759668.1 | 5 - 6 | 646 | 96 | ||
GenBank mRNA | BC098000.1 | - | 4 - 5 | 402 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |