Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109637.1 | NP_001103107.1 | 6 - 7 | 894 | 87 | |
XM_006233231.2 | XP_006233293.1 | 6 - 7 | 881 | 87 | ||
GenBank mRNA | BC158606.1 | - | 6 - 7 | 850 | 87 | |
FQ215425.1 | - | 6 - 7 | 835 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |