Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011986.1 | NP_001011986.1 | 2 - 3 | 210 | 64 | |
XM_006240885.2 | XP_006240947.1 | 1 - 2 | 167 | 64 | ||
XM_006240886.2 | XP_006240948.1 | 2 - 3 | 464 | 64 | ||
GenBank mRNA | BC079320.1 | - | 2 - 3 | 210 | 7135238 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |