Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109253.1 | NP_001102723.1 | 2 - 3 | 490 | 76 | |
GenBank mRNA | GU357481.1 | - | 2 - 3 | 205 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |