Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106628.1 | NP_001100098.1 | 5 - 6 | 542 | 115 | |
XM_006237676.1 | XP_006237738.1 | 5 - 6 | 547 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |