Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107903.1 | NP_001101373.1 | 6 - 7 | 985 | 118 | |
XM_006237827.2 | XP_006237889.1 | 6 - 7 | 1014 | 118 | ||
XM_006237832.2 | XP_006237894.1 | 6 - 7 | 1015 | 118 | ||
XM_008763539.1 | XP_008761761.1 | 6 - 7 | 1020 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |