Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013936.1 | NP_001013958.1 | 1 - 2 | 528 | 104 | |
GenBank mRNA | BC083723.1 | - | 1 - 2 | 528 | 7191003 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |