Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191063.1 | NP_001177992.1 | 9 - 10 | 1426 | 67 | |
XM_008764384.1 | XP_008762606.1 | 10 - 11 | 1590 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |