Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007650.1 | NP_001007651.1 | 1 - 2 | 268 | 123 | |
GenBank mRNA | AY346102.1 | - | 1 - 2 | 268 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |