Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025696.1 | NP_001020867.2 | 4 - 5 | 659 | 77 | |
GenBank mRNA | BC098795.1 | - | 4 - 5 | 659 | 7375769 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |