Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnali1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031647.1 | NP_001026817.1 | 2 - 3 | 225 | 84 | |
GenBank mRNA | BC099087.1 | - | 2 - 3 | 248 | 7455645 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |